ENA159843818 Enamine

2-chloro-1-(4-phenoxyphenyl)ethan-1-one

  • MDL number MFCD00997177
  • Cas Number : 13075-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33564
Availability details for
Quantity
  • Availability
  • Add to Cart