ENA159843823 Enamine

3-(3,4-dihydroisoquinolin-2(1H)-yl)propan-1-amine

  • MDL number MFCD06446877
  • Cas Number : 5596-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33580

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33580
Availability details for
Quantity
  • Availability
  • Add to Cart