ENA159843831 Enamine

2-chloro-N-[1-(4-chlorophenyl)propyl]acetamide

  • MDL number MFCD09971336
  • Cas Number : 40023-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28303

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28303
Availability details for
Quantity
  • Availability
  • Add to Cart