ENA159843833 Enamine

2-chloro-N-[cyclopropyl(4-methoxyphenyl)methyl]acetamide

  • MDL number MFCD00263892
  • Cas Number : 1031130-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28315
Availability details for
Quantity
  • Availability
  • Add to Cart