ENA159843836 Enamine

2-chloro-N-[cyclopropyl(4-methylphenyl)methyl]acetamide

  • MDL number MFCD09971337
  • Cas Number : 1031131-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28327
Availability details for
Quantity
  • Availability
  • Add to Cart