ENA159843857 Enamine

5-phenyl-1H-indole

  • MDL number MFCD06801971
  • Cas Number : 66616-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-73
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35207
Availability details for
Quantity
  • Availability
  • Add to Cart