ENA159843859 Enamine

2-{5-cyclopropyl-4-oxo-3H,4H-thieno[2,3-d]pyrimidin-2-yl}acetonitrile

  • MDL number MFCD09042124
  • Cas Number : 926199-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35209
Availability details for
Quantity
  • Availability
  • Add to Cart