ENA159843877 Enamine

1-(4-methoxy-3-phenylphenyl)ethan-1-one

  • MDL number MFCD09042414
  • Cas Number : 142500-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35457
Availability details for
Quantity
  • Availability
  • Add to Cart