ENA159843888 Enamine

7-methyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid

  • MDL number MFCD09439035
  • Cas Number : 102170-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35486
Availability details for
Quantity
  • Availability
  • Add to Cart