ENA159843898 Enamine

3-(pyridine-3-sulfonamido)propanoic acid

  • MDL number MFCD09933429
  • Cas Number : 1016775-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31720
Availability details for
Quantity
  • Availability
  • Add to Cart