ENA159843907 Enamine

(1E)-1-(4-nitrophenyl)ethanone phenylhydrazone

  • MDL number MFCD00436994
  • Cas Number : 77635-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31729

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31729
Availability details for
Quantity
  • Availability
  • Add to Cart