ENA159843908 Enamine

3-[4-(dimethylamino)phenyl]acrylonitrile

  • MDL number MFCD00001933
  • Cas Number : 32444-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165...168
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31730

Safety Information

signalword Warning
safety_statement 36
msds https://www.enaminestore.com/catalog/EN300-31730
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart