ENA159843914 Enamine

1-fluoro-4-trifluoromethanesulfonylbenzene

  • MDL number MFCD00474658
  • Cas Number : 944-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31735
Availability details for
Quantity
  • Availability
  • Add to Cart