ENA159843924 Enamine

N-(2-aminophenyl)benzamide

  • MDL number MFCD00034792
  • Cas Number : 721-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-152
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31745
Availability details for
Quantity
  • Availability
  • Add to Cart