ENA159843925 Enamine

N-(2-aminophenyl)-4-methylbenzamide

  • MDL number MFCD00447039
  • Cas Number : 71255-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31746
Availability details for
Quantity
  • Availability
  • Add to Cart