ENA159843926 Enamine

2-(4-aminophenoxy)-N,N-dimethylacetamide

  • MDL number MFCD09037319
  • Cas Number : 6271-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31747
Availability details for
Quantity
  • Availability
  • Add to Cart