ENA159843930 Enamine

2-piperazin-1-ylpyrazine

  • MDL number MFCD00040766
  • Cas Number : 34803-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48...50
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31751

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-31751
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart