ENA159843936 Enamine

2,5-dimethyl-1-(1,3-thiazol-2-yl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD09732323
  • Cas Number : 953902-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31756
Availability details for
Quantity
  • Availability
  • Add to Cart