ENA159843941 Enamine

1-(4-ethylbenzoyl)piperazine hydrochloride

  • MDL number MFCD09971555
  • Cas Number : 1171550-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 246...248
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31758

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31758
Availability details for
Quantity
  • Availability
  • Add to Cart