ENA159843942 Enamine

3-[(2-methyl-1H-imidazol-1-yl)methyl]benzonitrile

  • MDL number MFCD09931221
  • Cas Number : 1016716-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31759

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31759
Availability details for
Quantity
  • Availability
  • Add to Cart