ENA159843947 Enamine

2,5-dimethyl-1-(pyridin-3-yl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD09729672
  • Cas Number : 953717-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31763
Availability details for
Quantity
  • Availability
  • Add to Cart