ENA159843948 Enamine

4-fluorophthalic acid

  • MDL number MFCD06208213
  • Cas Number : 320-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151...153
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31765

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-31765
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart