ENA159843952 Enamine

1-[2-(methoxymethyl)phenyl]methanamine

  • MDL number MFCD09737485
  • Cas Number : 88032-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31770
Availability details for
Quantity
  • Availability
  • Add to Cart