ENA159843954 Enamine

2,5-dimethyl-1-[(pyridin-2-yl)methyl]-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD09733560
  • Cas Number : 953717-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31774
Availability details for
Quantity
  • Availability
  • Add to Cart