ENA159843956 Enamine

1-(3,5-dimethylbenzoyl)piperazine

  • MDL number MFCD08444699
  • Cas Number : 923163-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31776
Availability details for
Quantity
  • Availability
  • Add to Cart