ENA159843958 Enamine

4-(4-fluorophenoxy)butan-1-amine

  • MDL number MFCD07687074
  • Cas Number : 873790-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31778
Availability details for
Quantity
  • Availability
  • Add to Cart