ENA159843959 Enamine

2-(2,2,2-trifluoroethoxy)pyridine-3-carboxylic acid

  • MDL number MFCD00114457
  • Cas Number : 183368-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31779
Availability details for
Quantity
  • Availability
  • Add to Cart