ENA159843967 Enamine

2,5-dimethyl-1-(1-phenylethyl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD09732476
  • Cas Number : 953884-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31793
Availability details for
Quantity
  • Availability
  • Add to Cart