ENA159843986 Enamine

2-difluoromethanesulfonylaniline

  • MDL number MFCD00521859
  • Cas Number : 24906-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31819
Availability details for
Quantity
  • Availability
  • Add to Cart