ENA159843990 Enamine

3-methyl-1-[5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazol-5-amine

  • MDL number MFCD09047782
  • Cas Number : 926234-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31823
Availability details for
Quantity
  • Availability
  • Add to Cart