ENA159843998 Enamine

4-(ethoxymethyl)benzonitrile

  • MDL number MFCD09937541
  • Cas Number : 146781-27-3
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-31827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31827
Availability details for
Quantity
  • Availability
  • Add to Cart