ENA161173241 Enamine

2-{[bis(4-chlorophenyl)methoxy]methyl}oxirane

  • MDL number MFCD10686686
  • Cas Number : 1049605-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33600

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33600
Availability details for
Quantity
  • Availability
  • Add to Cart