ENA161173243 Enamine

3-(4-nitrophenyl)imidazo[1,5-a]pyrazin-1-ol hydrochloride

  • MDL number MFCD10686688
  • Cas Number : 1170599-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33660
Availability details for
Quantity
  • Availability
  • Add to Cart