ENA161173256 Enamine

6-chloro-5-nitro-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD10686703
  • Cas Number : 77859-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-35578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35578
Availability details for
Quantity
  • Availability
  • Add to Cart