ENA161173257 Enamine

2-chloro-4-phenylaniline hydrochloride

  • MDL number MFCD10686704
  • Cas Number : 1172521-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35579
Availability details for
Quantity
  • Availability
  • Add to Cart