ENA161173267 Enamine

dimethyl({[3-(4-nitrophenyl)-1,2,4-oxadiazol-5-yl]methyl})amine hydrochloride

  • MDL number MFCD10686714
  • Cas Number : 1170444-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-35595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35595
Availability details for
Quantity
  • Availability
  • Add to Cart