ENA161173268 Enamine

dimethyl({[3-(4-nitrophenyl)-1,2,4-oxadiazol-5-yl]methyl})amine

  • MDL number MFCD10686715
  • Cas Number : 1049873-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35598
Availability details for
Quantity
  • Availability
  • Add to Cart