ENA161173279 Enamine

methyl 1-(2-aminophenyl)-1H-1,2,3-triazole-4-carboxylate

  • MDL number MFCD10686731
  • Cas Number : 1049873-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35653
Availability details for
Quantity
  • Availability
  • Add to Cart