ENA161173293 Enamine

1-(heptafluoropropoxy)-4-nitrobenzene

  • MDL number MFCD11857765
  • Cas Number : 1146289-83-9
  • Molecular Weight : 307.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35740

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35740
Availability details for
Quantity
  • Availability
  • Add to Cart