ENA161173305 Enamine

[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]dimethylamine

  • MDL number MFCD10686765
  • Cas Number : 1049873-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-35791

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35791
Availability details for
Quantity
  • Availability
  • Add to Cart