ENA161173328 Enamine

2-(4-methoxybenzyl)-1H-benzimidazole

  • MDL number MFCD00729264
  • Cas Number : 53039-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33683

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33683
Availability details for
Quantity
  • Availability
  • Add to Cart