ENA161173344 Enamine

3-[(2-amino-1,3-thiazol-5-yl)methyl]benzonitrile

  • MDL number MFCD09051013
  • Cas Number : 937628-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-35185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35185
Availability details for
Quantity
  • Availability
  • Add to Cart