ENA161173347 Enamine

2-[5-amino-3-(2-methylphenyl)-1H-pyrazol-1-yl]ethan-1-ol

  • MDL number MFCD08445108
  • Cas Number : 956751-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35466
Availability details for
Quantity
  • Availability
  • Add to Cart