ENA161173369 Enamine

2-amino-4H,5H,6H,7H-thieno[2,3-c]pyridine-3-carbonitrile

  • MDL number MFCD08444198
  • Cas Number : 802025-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 247-249
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35538
Availability details for
Quantity
  • Availability
  • Add to Cart