ENA161173380 Enamine

methyl 2-hydroxy-5-(pyrrolidine-1-sulfonyl)benzoate

  • MDL number MFCD08142728
  • Cas Number : 893778-49-9
  • Molecular Weight : 285.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-29832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29832
Availability details for
Quantity
  • Availability
  • Add to Cart