ENA161173384 Enamine

8-nitro-1H,2H,4H,4aH,5H,6H-[1,4]oxazino[4,3-a]quinoline-5-carboxylic acid

  • MDL number MFCD10575753
  • Cas Number : 949270-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35565
Availability details for
Quantity
  • Availability
  • Add to Cart