ENA161173389 Enamine

1-nitro-4-(pentafluoroethoxy)benzene

  • MDL number MFCD02708322
  • Cas Number : 1743-96-0
  • Molecular Weight : 257.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35571
Availability details for
Quantity
  • Availability
  • Add to Cart