ENA161173396 Enamine

N-(3-aminophenyl)pyridine-4-carboxamide

  • MDL number MFCD00626063
  • Cas Number : 904013-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35582
Availability details for
Quantity
  • Availability
  • Add to Cart