ENA161173400 Enamine

2-(4-amino-2-chlorophenoxy)-N,N-diethylacetamide

  • MDL number MFCD09742296
  • Cas Number : 954571-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31744
Availability details for
Quantity
  • Availability
  • Add to Cart