ENA161173402 Enamine

4-[3-(1H-1,2,3,4-tetrazol-1-yl)phenyl]-1,3-thiazol-2-amine

  • MDL number MFCD09730146
  • Cas Number : 953754-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31782
Availability details for
Quantity
  • Availability
  • Add to Cart